Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008623.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008623.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008623.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008623.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008623.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008623.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159123.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159123.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159123.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159123.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159123.hg.1 |
TC0500008623.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500159120.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159120.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159120.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159120.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159120.hg.1 |
TC0500008623.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500159121.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159121.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159121.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159121.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159121.hg.1 |
TC0500008623.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008623.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008623.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008623.hg.1 |
ENST00000417667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008623.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008623.hg.1 |
NR_047470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008623.hg.1 |
NR_047470.1 |
Refseq |