Assay Details
Gene Symbol: NBPF13P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103466.1 | 4 - 5 | 519 | 99 | ||
GenBank mRNA | BX336604.2 | - | 4 - 5 | 519 | 99 | |
CB154168.1 | - | 2 - 3 | 196 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
RNA, variant U1 small nuclear 8 |
RNVU1-8 |
RNU1-145, vU1.8 |
-16.7kb | Sense | Chr. 1 - 147084616 - 147084756 on NCBI Build GRCh38 |
Downstream | protein kinase AMP-activated non-catalytic subunit beta 2 | PRKAB2 | 45.1kb | Sense | Chr. 1 - 147155106 - 147172589 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009765.hg.1 | ENST00000614199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009765.hg.1 | ENST00000614199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015681.hg.1 | ENST00000622818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015681.hg.1 | ENST00000622818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015681.hg.1 | ENST00000622818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009815.hg.1 | ENST00000450470.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_024442.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015621.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216933.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216933.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216933.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216933.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216933.hg.1 | TC0100015621.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100216931.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216931.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216931.hg.1 | TC0100015621.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100216938.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216938.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216938.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216938.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216938.hg.1 | TC0100015621.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009765.hg.1 | ENST00000614199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009765.hg.1 | ENST00000614199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009815.hg.1 | ENST00000450470.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015681.hg.1 | ENST00000622818.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015681.hg.1 | ENST00000622818.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015681.hg.1 | ENST00000622818.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216932.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216932.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216932.hg.1 | TC0100015621.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100216934.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216934.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216934.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216934.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216934.hg.1 | TC0100015621.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100216939.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216939.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216939.hg.1 | ENST00000444082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216939.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216939.hg.1 | TC0100015621.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100216940.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216940.hg.1 | NR_103466.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216940.hg.1 | TC0100015621.hg.1 | Clariom D TC |
