Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009433.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009433.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009433.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009433.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009433.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009433.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009433.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009433.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009433.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009433.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009433.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009433.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009433.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009433.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009433.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009433.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169731.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169731.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169731.hg.1 |
TC1400009433.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400169731.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169731.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169730.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169730.hg.1 |
NR_103769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169730.hg.1 |
TC1400009433.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400169730.hg.1 |
ENST00000531609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169730.hg.1 |
NR_103769.1 |
Refseq |