Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012074.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193932.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193932.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193932.hg.1 |
TC1200012074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200193932.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193932.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193933.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193933.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193933.hg.1 |
TC1200012074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200193933.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193933.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193936.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193936.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193936.hg.1 |
TC1200012074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200193936.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193936.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193934.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193934.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193934.hg.1 |
TC1200012074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200193934.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193934.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193935.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193935.hg.1 |
NR_103809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193935.hg.1 |
TC1200012074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200193935.hg.1 |
ENST00000548738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193935.hg.1 |
NR_103809.1 |
Refseq |