Assay Details
Gene Symbol: NRG1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104156.1 | 3 - 4 | 333 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Werner syndrome RecQ like helicase | WRN |
RECQ3, RECQL2, RECQL3 |
-856.6kb | Sense | Chr. 8 - 31033262 - 31173761 on NCBI Build GRCh38 |
Downstream | NRG1 intronic transcript 3 | NRG1-IT3 | 301.3kb | Sense | Chr. 8 - 32440746 - 32442725 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
