Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000523743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000523743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000523743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000523743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000523785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000523785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
ENST00000523785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007225.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149727.hg.1 |
ENST00000523785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149727.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149727.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149727.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149727.hg.1 |
TC0800007225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800149727.hg.1 |
ENST00000523785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149727.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149727.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149727.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149711.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149711.hg.1 |
TC0800007225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800149711.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000520444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000521463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000523743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000523743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000523743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000523743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000523785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000523785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
ENST00000523785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007225.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
ENST00000523743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
ENST00000523785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
NR_104158.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
TC0800007225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
ENST00000523743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
ENST00000523785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
NR_104156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
NR_104157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149726.hg.1 |
NR_104158.1 |
Refseq |