Assay Details
Gene Symbol: LOC101928336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110396.1 | 3 | 316 | 65 | ||
GenBank mRNA | BC041420.1 | - | 3 | 316 | 65 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BE778080.1 | - | 295 | Chr. - - [-] on Build | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | progesterone receptor membrane component 1 | PGRMC1 |
HPR6.6, MPR |
-81.7kb | Sense | Chr. X - 119236245 - 119244466 on NCBI Build GRCh38 |
Downstream | solute carrier family 25 member 43 | SLC25A43 | 73.0kb | Sense | Chr. X - 119399295 - 119454478 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
