Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017465.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232109.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232109.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232109.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232109.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232109.hg.1 |
TC0100017465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100232109.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232109.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232109.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232109.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232112.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232112.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232112.hg.1 |
TC0100017465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100232112.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232112.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232108.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232108.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232108.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232108.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232108.hg.1 |
TC0100017465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100232108.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232108.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232108.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232108.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
ENST00000436706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
ENST00000453760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
NR_110669.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017465.hg.1 |
NR_110669.1 |
Refseq |