Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173847.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173847.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173847.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173847.hg.1 |
TC0800010878.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800173847.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173847.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173847.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
TC0800010878.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173851.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173848.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173848.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173848.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173848.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173848.hg.1 |
TC0800010878.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800173848.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173848.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173848.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173848.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
NR_125389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
TC0800010878.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
ENST00000519242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
ENST00000522302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
ENST00000522807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173850.hg.1 |
NR_125389.1 |
Refseq |