Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007889.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007889.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007889.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007889.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007889.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007889.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154093.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154093.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154093.hg.1 |
TC0400007889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400154093.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154093.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007889.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007889.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007889.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007889.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007889.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007889.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154094.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154094.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154094.hg.1 |
TC0400007889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400154094.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154094.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154092.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154092.hg.1 |
NR_125906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154092.hg.1 |
TC0400007889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400154092.hg.1 |
ENST00000501239.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154092.hg.1 |
NR_125906.1 |
Refseq |