Assay Details
Gene Symbol: LOC729218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103825.1 | 3 | 1374 | 73 | ||
NR_109983.1 | 10 | 2317 | 73 | |||
GenBank mRNA | AK024248.1 | - | 10 | 2288 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | centrosomal protein 170 pseudogene 1 | CEP170P1 |
CEP170L, FAM68B, KIAA0470L |
-76.2kb | Sense | Chr. 4 - 118516340 - 118554204 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101929741 | LOC101929741 | 29.1kb | Anti-sense | Chr. 4 - 118664087 - 118685341 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000411293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011073.hg.1 | ENST00000444843.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011073.hg.1 | ENST00000446669.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158607.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158607.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158607.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158607.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158607.hg.1 | TC0400008541.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400158607.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158607.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158607.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158607.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000411293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008541.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158612.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158612.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158612.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158612.hg.1 | TC0400008541.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400158612.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158612.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158612.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011073.hg.1 | ENST00000444843.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011073.hg.1 | ENST00000446669.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158606.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158606.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158606.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158606.hg.1 | TC0400008541.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400158606.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158606.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158606.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158613.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158613.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158613.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158613.hg.1 | TC0400008541.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400158613.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158613.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158613.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158615.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158615.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158615.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158615.hg.1 | TC0400008541.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400158615.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158615.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158615.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158611.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158611.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158611.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158611.hg.1 | TC0400008541.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400158611.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158611.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158611.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158614.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158614.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158614.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158614.hg.1 | TC0400008541.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400158614.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158614.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158614.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158616.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158616.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158616.hg.1 | TC0400008541.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400158616.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158616.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158608.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158608.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158608.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158608.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158608.hg.1 | TC0400008541.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400158608.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158608.hg.1 | ENST00000567913.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158608.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158608.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158610.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158610.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158610.hg.1 | NR_109983.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158610.hg.1 | TC0400008541.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400158610.hg.1 | ENST00000426314.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158610.hg.1 | NR_103825.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158610.hg.1 | NR_109983.1 | Refseq |
