Assay Details
Gene Symbol: CCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164680.1 | NP_001158152.1 | 2 - 3 | 218 | 108 | |
GenBank mRNA | AF262303.1 | - | 2 - 3 | 138 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF262303.1, NM_001164680.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of endothelial cell proliferation
chemotaxis
inflammatory response
cellular defense response
cell adhesion
adenylate cyclase-modulating G protein-coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
viral process
positive regulation of angiogenesis
chemokine-mediated signaling pathway
chemokine receptor activity
protein binding
C-C chemokine receptor activity
positive regulation of endothelial cell proliferation
chemotaxis
inflammatory response
cellular defense response
cell adhesion
adenylate cyclase-modulating G protein-coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
viral process
positive regulation of angiogenesis
chemokine-mediated signaling pathway
chemokine receptor activity
protein binding
C-C chemokine receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
