Assay Details
Gene Symbol: PPP4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318028.1 | NP_001304957.1 | 2 - 3 | 368 | 72 | |
GenBank mRNA | BX089727.1 | - | 2 - 3 | 177 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
