Assay Details
Gene Symbol: XLOC_008559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131214.1 | 1 - 2 | 105 | 138 | ||
GenBank mRNA | HY074184.1 | - | 1 - 2 | 105 | 138 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ankyrin repeat domain 1 | ANKRD1 |
ALRP, C-193, CARP, CVARP, MCARP, bA320F15.2 |
-26.0kb | Anti-sense | Chr. 10 - 90912100 - 90921275 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC105378430 | LOC105378430 | 85.5kb | Sense | Chr. 10 - 91033166 - 91041255 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
