Assay Details
Gene Symbol: CRAT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131985.1 | 1 - 2 | 179 | 82 | ||
GenBank mRNA | DA425353.1 | - | 1 - 2 | 179 | 82 | |
DA428755.1 | - | 1 - 2 | 128 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 508 | LINC00508 | -42.0kb | Sense | Chr. 12 - 127883989 - 127983891 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC100996679 | LOC100996679 | 59.8kb | Sense | Chr. 12 - 128086982 - 128118141 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
