Assay Details
Gene Symbol: LOC727993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134520.1 | 1 - 2 | 441 | 85 | ||
GenBank mRNA | AK090681.1 | - | 1 - 2 | 408 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | signal regulatory protein alpha | SIRPA |
BIT, CD172A, MFR, MYD-1, P84, PTPNS1, SHPS1, SIRP |
-7.3kb | Sense | Chr. 20 - 1894117 - 1939895 on NCBI Build GRCh38 |
Downstream | prodynorphin | PDYN |
ADCA, PENKB, SCA23 |
27.2kb | Anti-sense | Chr. 20 - 1978756 - 1994285 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
