Assay Details
Gene Symbol: LOC101929134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134663.1 | 1 - 2 | 129 | 115 | ||
GenBank mRNA | BX106511.1 | - | 1 - 2 | 113 | 115 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PIGY upstream reading frame | PYURF | PREY | -9.0 | Anti-sense | Chr. 4 - 88520978 - 88523801 on NCBI Build GRCh38 |
Downstream | HECT and RLD domain containing E3 ubiquitin protein ligase 3 | HERC3 | 67.4kb | Sense | Chr. 4 - 88592423 - 88709303 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
