Assay Details
Gene Symbol: LOC105378933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135035.1 | 1 - 2 | 112 | 102 | ||
GenBank mRNA | BX101087.1 | - | 1 - 2 | 99 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | T-box 15 | TBX15 | TBX14 | -10.8kb | Sense | Chr. 1 - 118883043 - 118989556 on NCBI Build GRCh38 |
Downstream |
tryptophanyl tRNA synthetase 2, mitochondrial |
WARS2 | TrpRS | 29.8kb | Sense | Chr. 1 - 119031216 - 119140701 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
