Assay Details
Gene Symbol: LOC105378663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135048.1 | 1 - 2 | 179 | 92 | ||
GenBank mRNA | BX115242.1 | - | 1 - 2 | 130 | 92 | |
HY234287.1 | - | 1 - 2 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GJA9-MYCBP readthrough | GJA9-MYCBP | Anti-sense | Chr. 1 - 38862490 - 38881626 on NCBI Build GRCh38 | ||
Downstream | rhomboid like 2 | RHBDL2 | RRP2 | 9.6kb | Anti-sense | Chr. 1 - 38885806 - 38941830 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
