Assay Details
Gene Symbol: SEMA3F-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135301.1 | 1 - 2 | 354 | 83 | ||
GenBank mRNA | AK125500.1 | - | 1 - 2 | 354 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | basic proline-rich protein-like | LOC107986035 | Anti-sense | Chr. 3 - 50154670 - 50155789 on NCBI Build GRCh38 | ||
Downstream | microRNA 566 | MIR566 |
MIRN566, hsa-mir-566 |
17.2kb | Anti-sense | Chr. 3 - 50173326 - 50173419 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
