Assay Details
Gene Symbol: SEMA3F-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135301.1 | 2 - 3 | 468 | 81 | ||
GenBank mRNA | AK125500.1 | - | 2 - 3 | 468 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RNA binding motif protein 5 | RBM5 |
G15, H37, LUCA15, RMB5 |
-3.6kb | Anti-sense | Chr. 3 - 50088908 - 50118964 on NCBI Build GRCh38 |
Downstream | basic proline-rich protein-like | LOC107986035 | 18.1kb | Anti-sense | Chr. 3 - 50154670 - 50155789 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
