Assay Details
Gene Symbol: LINC00484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135306.1 | 2 - 3 | 162 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100129316 | LOC100129316 | -44.1kb | Anti-sense | Chr. 9 - 91063294 - 91075132 on NCBI Build GRCh38 | |
Downstream | AU RNA binding protein/enoyl-CoA hydratase | AUH | 31.1kb | Anti-sense | Chr. 9 - 91213815 - 91361969 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000414768.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000414768.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000414768.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000414768.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000414768.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000421595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000421595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000421595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000439438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000439438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000439438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000439438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000439438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000588550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000588550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000588550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000588550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000608240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000608240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000608240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000609752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000609752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000609752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000609752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007919.hg.1 | ENST00000609752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000414768.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000414768.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000414768.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000414768.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000414768.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000421595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000421595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000421595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000423719.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000437389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000439438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000439438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000439438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000439438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000439438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000588550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000588550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000588550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000588550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000608240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000608240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000608240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000609752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000609752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000609752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000609752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007919.hg.1 | ENST00000609752.1 | Ensembl |
