Assay Details
Gene Symbol: LOC105371814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135674.1 | 2 - 3 | 475 | 55 | ||
GenBank mRNA | CX873301.1 | - | 2 - 3 | 478 | 55 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | calcium binding and coiled-coil domain 2 | CALCOCO2 | NDP52 | -14.0kb | Anti-sense | Chr. 17 - 48830988 - 48865245 on NCBI Build GRCh38 |
Downstream |
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9) |
ATP5G1 |
ATP5A, ATP5G |
109.0 | Anti-sense | Chr. 17 - 48892786 - 48895871 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
