Assay Details
Gene Symbol: LOC101060391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_136151.1 | 1 - 2 | 105 | 59 | ||
GenBank mRNA | DB544287.1 | - | 1 - 2 | 93 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101060385 | LOC101060385 | -44.5kb | Sense | Chr. 2 - 900216 - 905451 on NCBI Build GRCh38 | |
Downstream | thyroid peroxidase | TPO |
MSA, TDH2A, TPX |
462.1kb | Anti-sense | Chr. 2 - 1413461 - 1542729 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
