Assay Details
Gene Symbol: MIR28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029502.1 | 1 | 44 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | LPP antisense RNA 1 | LPP-AS1 | -120.1kb | Anti-sense | Chr. 3 - 188562237 - 188568666 on NCBI Build GRCh38 | |
Downstream | TPRG1 antisense RNA 1 | TPRG1-AS1 | 252.8kb | Anti-sense | Chr. 3 - 188941715 - 188947639 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
