Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000583962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000583962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
ENST00000583962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
NR_104185.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
NR_104185.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
NR_104185.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012212.hg.1 |
NR_104185.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000577537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000582324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000583962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000583962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
ENST00000583962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
NR_104185.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
NR_104185.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
NR_104185.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012212.hg.1 |
NR_104185.1 |
Refseq |