Assay Details
Gene Symbol: MIR6834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_106892.1 | 1 | 43 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | prefoldin subunit 6 | PFDN6 |
H2-KE2, HKE2, KE-2, PFD6 |
Sense | Chr. 6 - 33289597 - 33290934 on NCBI Build GRCh38 | |
Downstream | ral guanine nucleotide dissociation stimulator like 2 | RGL2 |
HKE1.5, KE1.5, RAB2L |
1.3kb | Anti-sense | Chr. 6 - 33291654 - 33299388 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374610.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374610.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374610.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374610.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000374610.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000463584.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000463584.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000463584.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000463584.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000463584.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | ENST00000622807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007695.hg.1 | NR_106892.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374606.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374610.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374610.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374610.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374610.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000374610.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395131.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000395134.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000463584.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000463584.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000463584.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000463584.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000463584.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000491382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | ENST00000622807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001185181.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265595.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_001265596.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NM_014260.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007695.hg.1 | NR_106892.1 | Refseq |
