Assay Details
Gene Symbol: PCAT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109832.1 | 3 - 4 | 381 | 62 | ||
GenBank mRNA | BG248682.1 | - | 3 - 4 | 382 | 62 | |
HQ605085.1 | - | 3 - 4 | 397 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000629869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000629869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | ENST00000629869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006848.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147203.hg.1 | TC2200006848.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147203.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147203.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147203.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147203.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147202.hg.1 | TC2200006848.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147202.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147202.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147202.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147202.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147202.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147202.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147195.hg.1 | TC2200006848.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147195.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147195.hg.1 | ENST00000629869.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147195.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147195.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147195.hg.1 | ENST00000629869.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147195.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147199.hg.1 | TC2200006848.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147199.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147199.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147199.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147199.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147199.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147199.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000626825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000627127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000629869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000629869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | ENST00000629869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006848.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147194.hg.1 | TC2200006848.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147194.hg.1 | NR_109832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147194.hg.1 | NR_109832.1 | Refseq |
