Assay Details
Gene Symbol: LOC101927348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110915.1 | 2 - 3 | 300 | 84 | ||
GenBank mRNA | BC039386.1 | - | 2 - 3 | 300 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927311 | LOC101927311 | -2.9kb | Sense | Chr. 16 - 14009276 - 14016018 on NCBI Build GRCh38 | |
Downstream | MKL1/myocardin like 2 | MKL2 |
MRTF-B, MRTFB, NPD001 |
50.6kb | Anti-sense | Chr. 16 - 14071076 - 14266779 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
