Assay Details
Gene Symbol: CHN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120524.1 | 1 - 2 | 511 | 83 | ||
GenBank mRNA | EU732750.1 | - | 1 - 2 | 112 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC102724484 | LOC102724484 | -270.7kb | Anti-sense | Chr. 7 - 29199040 - 29208970 on NCBI Build GRCh38 | |
Downstream | proline rich 15 | PRR15 | 63.8kb | Sense | Chr. 7 - 29563796 - 29567300 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
