Assay Details
Gene Symbol: LINC01044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126345.1 | 2 - 3 | 472 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1043 | LINC01043 | -801.0 | Anti-sense | Chr. 13 - 112313467 - 112321766 on NCBI Build GRCh38 | |
Downstream | sperm acrosome associated 7 | SPACA7 | C13orf28 | 48.8kb | Anti-sense | Chr. 13 - 112376300 - 112434695 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
