Assay Details
Gene Symbol: LINC01195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126349.1 | 3 - 4 | 330 | 89 | ||
GenBank mRNA | CR748468.1 | - | 3 - 4 | 329 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1177 | LINC01177 | -1.2kb | Sense | Chr. 16 - 9441294 - 9444985 on NCBI Build GRCh38 | |
Downstream | glutamate ionotropic receptor NMDA type subunit 2A | GRIN2A |
EPND, FESD, GluN2A, LKS, NMDAR2A, NR2A |
305.4kb | Sense | Chr. 16 - 9753404 - 10183364 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
