Assay Details
Gene Symbol: LINC01177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126397.1 | 3 - 4 | 315 | 83 | ||
GenBank mRNA | AA725682.1 | - | 1 - 2 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927009 | LOC101927009 | -33.2kb | Anti-sense | Chr. 16 - 9355588 - 9408093 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 1195 | LINC01195 | 3.4kb | Sense | Chr. 16 - 9446013 - 9455505 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
