Assay Details
Gene Symbol: STX18-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126434.1 | 2 - 3 | 405 | 99 | ||
GenBank mRNA | DB060204.1 | - | 2 - 3 | 406 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | syntaxin 18 | STX18 | Ufe1 | Sense | Chr. 4 - 4418968 - 4542338 on NCBI Build GRCh38 | |
Downstream | STX18 antisense RNA 1 (head to head) | STX18-AS1 | 63.2kb | Anti-sense | Chr. 4 - 4542131 - 4710938 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
