Assay Details
Gene Symbol: ENPP7P13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_130771.1 | 1 - 2 | 296 | 73 | ||
GenBank mRNA | JF934746.1 | - | 1 - 2 | 296 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105379474 | LOC105379474 | -282.9kb | Anti-sense | Chr. 16 - 33484765 - 33486497 on NCBI Build GRCh38 | |
Downstream | immunoglobulin heavy variable 3/OR16-7 (pseudogene) | IGHV3OR16-7 |
IGH, IGHV, IGHV3/OR16-7, IGHV3OR167 |
160.7kb | Anti-sense | Chr. 16 - 33938339 - 33938639 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
