Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000424701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000424701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000426752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000426752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000426752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000452286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000452286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000452286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000452286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008824.hg.1 |
ENST00000452286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000424701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000424701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000426752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000426752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000426752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000449761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000452286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000452286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000452286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000452286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008824.hg.1 |
ENST00000452286.1 |
Ensembl |