Assay Details
Gene Symbol: DANT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_130730.1 | 1 | 101 | 82 | ||
NR_132337.1 | 1 | 101 | 82 | |||
GenBank mRNA | BC003645.1 | - | 1 | 79 | 82 | |
BE297956.1 | - | 1 | 102 | 82 | ||
BM925596.1 | - | 1 | 123 | 82 | ||
BX642309.1 | - | 1 | 108 | 82 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | DA521463.1 | - | 99 | Chr. - - [-] on Build | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | plastin 3 | PLS3 |
BMND18, T-plastin |
-318.1kb | Anti-sense | Chr. X - 115560850 - 115650861 on NCBI Build GRCh38 |
Downstream | angiotensin II receptor type 2 | AGTR2 |
AT2, ATGR2, MRX88 |
201.6kb | Anti-sense | Chr. X - 116170705 - 116174972 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
