Assay Details
Gene Symbol: LOC105374389 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_136317.1 | 2 | 313 | 98 | ||
GenBank mRNA | AA448482.1 | - | 1 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CAP-Gly domain containing linker protein family member 4 | CLIP4 | RSNL2 | -290.9kb | Anti-sense | Chr. 2 - 29097676 - 29183813 on NCBI Build GRCh38 |
Downstream | yippee like 5 | YPEL5 | CGI-127 | 671.5kb | Anti-sense | Chr. 2 - 30146568 - 30160533 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
