Assay Details
Gene Symbol: C16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011522462.2 | XP_011520764.1 | 1 | 117 | 104 | |
GenBank mRNA | BI831003.1 | - | 1 | 121 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011522462.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
