Assay Details
Gene Symbol: TRIM16L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005256480.4 | XP_005256537.1 | 2 - 3 | 236 | 61 | |
XM_005256481.4 | XP_005256538.1 | 2 - 3 | 236 | 61 | ||
XM_011523690.2 | XP_011521992.1 | 2 - 3 | 236 | 61 | ||
XM_017024245.1 | XP_016879734.1 | 2 - 3 | 236 | 61 | ||
XM_017024246.1 | XP_016879735.1 | 2 - 3 | 236 | 61 | ||
XM_017024247.1 | XP_016879736.1 | 2 - 3 | 236 | 61 | ||
GenBank mRNA | AK293428.1 | - | 2 - 3 | 236 | 61 | |
AL080124.1 | - | 1 - 2 | 97 | 61 | ||
BC106034.1 | - | 2 - 3 | 206 | 61 | ||
BC127827.1 | - | 1 - 2 | 58 | 40131713 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK293428.1, BC127827.1, XM_005256480.4, XM_005256481.4, XM_011523690.2, XM_017024245.1, XM_017024246.1, XM_017024247.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
