Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000449405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000449405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000449405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
ENST00000449405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000445279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000449405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000449405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000449405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
ENST00000449405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011478.hg.1 |
NR_110179.1 |
Refseq |