Assay Details
Gene Symbol: LOC646743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY726562.1 | - | 4 - 5 | 614 | 80 |
Gene Symbol: LOC107985778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017005439.1 | XP_016860928.1 | 2 - 3 | 618 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
