Assay Details
Gene Symbol: LOC102724849 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110607.1 | 3 - 4 | 563 | 110 | ||
GenBank mRNA | BX095234.1 | - | 3 - 4 | 563 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
