Assay Details
Gene Symbol: HK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000189.4 | NP_000180.2 | 1 | 1437 | 89 | |
GenBank mRNA | AW134604.1 | - | 1 | 111 | 89 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BC013295.1 | - | 1 | Chr. 2 - 74833399 - 74835160 [-] on Build GRCh38 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_000189.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cellular glucose homeostasis
response to ischemia
glycolytic process
lactation
apoptotic mitochondrial changes
negative regulation of mitochondrial membrane permeability
regulation of glucose import
carbohydrate phosphorylation
glucose 6-phosphate metabolic process
canonical glycolysis
establishment of protein localization to mitochondrion
maintenance of protein location in mitochondrion
glucose transmembrane transport
positive regulation of autophagy of mitochondrion in response to mitochondrial depolarization
negative regulation of reactive oxygen species metabolic process
glucokinase activity
hexokinase activity
protein binding
ATP binding
glucose binding
fructokinase activity
mannokinase activity
cellular glucose homeostasis
response to ischemia
glycolytic process
lactation
apoptotic mitochondrial changes
negative regulation of mitochondrial membrane permeability
regulation of glucose import
carbohydrate phosphorylation
glucose 6-phosphate metabolic process
canonical glycolysis
establishment of protein localization to mitochondrion
maintenance of protein location in mitochondrion
glucose transmembrane transport
positive regulation of autophagy of mitochondrion in response to mitochondrial depolarization
negative regulation of reactive oxygen species metabolic process
glucokinase activity
hexokinase activity
protein binding
ATP binding
glucose binding
fructokinase activity
mannokinase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
