Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |