Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000304552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000304552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000304552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000304552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000438735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000438735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000438735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000438735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000457814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000457814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000458629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000458629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000458629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000458629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
ENST00000458629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
NM_006564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
NM_006564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
NM_006564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007251.hg.1 |
NM_006564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000304552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000304552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000304552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000304552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000438735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000438735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000438735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000438735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000457814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000457814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000458629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000458629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000458629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000458629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
ENST00000458629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
NM_006564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
NM_006564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
NM_006564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007251.hg.1 |
NM_006564.1 |
Refseq |