Assay Details
Gene Symbol: SUCLG2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109993.1 | 3 - 4 | 624 | 92 | ||
GenBank mRNA | BC038210.1 | - | 3 - 4 | 624 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000464420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000482677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | ENST00000496640.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007728.hg.1 | NR_109993.1 | Refseq |
