Assay Details
Gene Symbol: EHHADH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038990.1 | 2 - 3 | 207 | 73 | ||
GenBank mRNA | AL703608.1 | - | 2 - 3 | 425 | 74 | |
BC047598.1 | - | 2 - 3 | 207 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000417720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | ENST00000444571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | NR_038990.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009730.hg.1 | NR_038990.1 | Refseq |
