Assay Details
Gene Symbol: SEC22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012430.4 | NP_036562.2 | 6 - 7 | 800 | 77 | |
XM_011512673.2 | XP_011510975.1 | 6 - 7 | 2316 | 77 | ||
XM_011512675.2 | XP_011510977.1 | 7 - 8 | 967 | 77 | ||
XM_011512676.2 | XP_011510978.1 | 6 - 7 | 856 | 77 | ||
GenBank mRNA | AF100749.1 | - | 6 - 7 | 782 | 77 | |
AK057587.1 | - | 5 - 6 | 1616 | 77 | ||
AK315771.1 | - | 6 - 7 | 769 | 77 | ||
BC007122.1 | - | 5 - 6 | 748 | 3459649 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000309934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000466950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000473494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000477063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000480631.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000481965.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000487572.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000491366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | ENST00000492595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013865.hg.1 | NM_012430.4 | Refseq |
