Assay Details
Gene Symbol: PCDH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011513844.2 | XP_011512146.1 | 4 - 5 | 4596 | 78 | |
XM_017008264.1 | XP_016863753.1 | 6 - 7 | 4660 | 78 | ||
XM_017008272.1 | XP_016863761.1 | 3 - 4 | 4535 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000507864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000507864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000507864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000509925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000509925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000509925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000509925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000610830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000610830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000610830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000361762.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000507864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000507864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000507864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000509759.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000509925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000509925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000509925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000509925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000511884.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000543491.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000610830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000610830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000610830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | ENST00000621961.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_001173523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_002589.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007169.hg.1 | NM_032457.3 | Refseq |
